Anti-p53 Rabbit Polyclonal Antibody

Supplier: Antibodies.com LLC
A95479-100
77050-464EA 473.8 USD
77050-464
Anti-p53 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to p53 for WB, IHC, IF and ELISA with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IHC, IF, ELISA
  • Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, IF: 1:100-1:500, ELISA: 1:10000

Type: Primary
Antigen: p53
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR