Anti-CD63 Mouse Monoclonal Antibody [Clone: MEM-259] (Biotin)

Supplier: Antibodies.com LLC
A85980-100
76957-354EA 391.4 USD
76957-354
Anti-CD63 Mouse Monoclonal Antibody [Clone: MEM-259] (Biotin)
Antibodies
Mouse monoclonal [MEM-259] antibody to CD63 (Biotin) for Flow Cytometry, IP, IHC-P and ICC with samples derived from Human.

  • Validated Applications: Flow Cytometry, IP, IHC-P, ICC
  • Recommended Dilutions: Flow Cytometry: 1-3 µg/ml

Type: Primary
Antigen: CD63
Clonality: Monoclonal
Clone: MEM-259
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR