Anti-CD52 Mouse Monoclonal Antibody [Clone: HI186] (PE (Phycoerythrin))

Supplier: Antibodies.com LLC
A85764-100
76952-404EA 430 USD
76952-404
Anti-CD52 Mouse Monoclonal Antibody [Clone: HI186] (PE (Phycoerythrin))
Antibodies
Mouse monoclonal [HI186] antibody to CD52 (PE) for Flow Cytometry with samples derived from Human.

  • Validated Applications: Flow Cytometry
  • Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD52
Clonality: Monoclonal
Clone: HI186
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR