Anti-Cathelicidin/CLP Rabbit Polyclonal Antibody

Supplier: Antibodies.com LLC
A93041-50 A93041-100
77213-204EA 225 USD
77213-204 77201-198
Anti-Cathelicidin/CLP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Cathelicidin/CLP for WB, IHC and ICC/IF with samples derived from Human and Rat.

  • Validated Applications: WB, IHC, ICC/IF
  • Recommended Dilutions: WB: 1:500-1:2, 000, IHC: 1:50-1:200, ICC/IF: 1:50-1:200

Type: Primary
Antigen: Cathelicidin/CLP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR