Accessories for Endotoxin Test Related Products
Supplier: Fujifilm
FUJIFILM Wako offers a premiere collection of endotoxin-free accessories including test tubes, caps and LAL reagent water designed for worry-free endotoxin testing.
Lysate reagent water: Endotoxin-free water (<0.001 EU/ml), Non-LAL reactive, Used to perform dilution series of RSE and CSE, Steam-sterilized by USP standards, Great for reconstitution of LAL, RSE and CSE, Used for sample dilutions.
Gel clot reaction tubes: Endotoxin-free test tubes (<0.001 EU/tube), Endotoxin-free tubes depyrogenated at 250 °C, Made with borosilicate glass and designed for gel clot testing.
Limulus test tube S: Endotoxin-free test tubes (<0.001 EU/tube), Designed for gel clot, KTA and KCA testing on the Toxinometer®, Made with borosilicate glass.
Depyrogenated dilution tubes: Endotoxin-free test tubes (<0.001 EU/tube), Made with borosilicate glass. Aluminum caps: endotoxin free and made to fit reaction and dilution tubes.
Endotoxin free and suitable to use with all LAL testing methodologies.