Anti-TRP1 Mouse Monoclonal Antibody [clone: TYRP1/807]

Supplier: Antibodies.com LLC
A250254-100
76968-626EA 525 USD
76968-626
Anti-TRP1 Mouse Monoclonal Antibody [clone: TYRP1/807]
Antibodies
Mouse monoclonal [TYRP1/807] antibody to TRP1 for IHC-P with samples derived from Human and Mouse.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 3 to 6 µg/ml

Type: Primary
Antigen: TRP1
Clonality: Monoclonal
Clone: TYRP1/807
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR