Anti-CD3 Mouse Monoclonal Antibody [clone: OKT3] (APC (Allophycocyanin))

Supplier: Antibodies.com LLC
A121901-100
76963-740EA 430 USD
76963-740
Anti-CD3 Mouse Monoclonal Antibody [clone: OKT3] (APC (Allophycocyanin))
Antibodies
Mouse monoclonal [OKT3] antibody to CD3 (APC) for Flow Cytometry with samples derived from Human.

  • Validated Applications: Flow Cytometry
  • Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: OKT3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR