Anti-COL28A1 Rabbit Polyclonal Antibody

Supplier: Antibodies.com LLC
A98612-100
77072-200EA 473.8 USD
77072-200
Anti-COL28A1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to COL28A1 for WB and ELISA with samples derived from Human and Mouse.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:2000, ELISA: 1:10000

Type: Primary
Antigen: COL28A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR