4-Iodobenzonitrile 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
p-Iodobenzonitrile
A986300-1000G
A986300-100G
A986300-10G
A986300-25G
A986300-500G
A986300-5G
77282-118EA
679.8
USD
77282-118
77282-120
77282-122
77282-124
77282-126
77282-128
4-Iodobenzonitrile 98%
4-Iodobenzonitrile
Caution: for Research Use Only
Formula:
C₇H₄IN MW: 229.02 g/mol |
MDL Number:
MFCD00051310 CAS Number: 3058-39-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...