1,3-Dichloro-4-fluorobenzene 98%

Supplier: Ambeed
Danger

Synonyms: 2,4-Dichlorofluorobenzene, 2,4-Dichloro-1-fluorobenzene

A961183-10G A961183-25G A961183-5G
77256-854EA 6 USD
77256-854 77256-856 77256-858
1,3-Dichloro-4-fluorobenzene 98%
1,3-Dichloro-4-fluorobenzene

Caution: for Research Use Only

Formula: C₆H₃Cl₂F
MDL Number: MFCD00000580
CAS Number: 1435-48-9
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR