2-Cyclopentylethanol 98%

Supplier: Ambeed
Warning

A927771-1G A927771-25G A927771-5G
77268-942EA 7 USD
77268-942 77268-944 77268-946
2-Cyclopentylethanol 98%
2-Cyclopentylethanol

Caution: for Research Use Only

Formula: C₇H₁₄O
MW: 114.19 g/mol
MDL Number: MFCD00019301
CAS Number: 766-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR