2',7'-Dichlorofluorescein 95%

Supplier: Ambeed
Warning

A864353-100G A864353-1G A864353-5G
77256-386EA 322 USD
77256-386 77256-388 77256-390
2',7'-Dichlorofluorescein 95%
2',7'-Dichlorofluorescein

Caution: for Research Use Only

Formula: C₂₀H₁₀Cl₂O₅
MDL Number: MFCD00005047
CAS Number: 76-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR