1-Tridecene 97% + stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A740122-1G A740122-25G A740122-5G
77243-846EA 15 USD
77243-846 77243-848 77243-850
1-Tridecene 97% + stabilized
1-Tridecene

Caution: for Research Use Only

Formula: C₁₃H₂₆
MDL Number: MFCD00008976
CAS Number: 2437-56-1
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR