1-Phenyl-1,3-butandione 97%

Supplier: Ambeed
Warning

Synonyms: 1-Phenylbutane-1,3-dione, 1-Benzoylacetone, Benzoylacetone, 1-Phenyl-1,3-butanedione

A726729-100G A726729-25G A726729-500G A726729-5G
77243-740EA 85 USD
77243-740 77243-742 77243-744 77243-746
1-Phenyl-1,3-butandione 97%
1-Phenyl-1,3-butandione

Caution: for Research Use Only

Formula: C₆H₅C(O)CH₂C(O)CH₃
MW: 162.19 g/mol
Boiling Pt: 262
MDL Number: MFCD00008786
CAS Number: 93-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR