Ethyl-3-amino-3-iminopropanoate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-amidinoacetate hydrochloride

A720723-100G A720723-1G A720723-250MG A720723-25G A720723-5G
77280-960EA 349 USD
77280-960 77280-962 77280-964 77280-966 77280-968
Ethyl-3-amino-3-iminopropanoate hydrochloride 97%
Ethyl-3-amino-3-iminopropanoate hydrochloride

Caution: for Research Use Only

Formula: C₅H₁₁ClN₂O₂
MW: 166.60 g/mol
MDL Number: MFCD06797615
CAS Number: 57508-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR