4-(1-Adamantyl)phenol 97%

Supplier: Ambeed
Warning

A698501-100G A698501-10G A698501-1G A698501-250MG A698501-25G A698501-5G
77293-268EA 262.99 USD
77293-268 77293-270 77293-272 77293-274 77293-276 77293-278
4-(1-Adamantyl)phenol 97%
4-(1-Adamantyl)phenol

Caution: for Research Use Only

Formula: C₁₆H₂₀O
MDL Number: MFCD00168143
CAS Number: 29799-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR