3-Amino-4-phenyl-1H-pyrazole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Phenyl-1H-pyrazol-3-amine

A695258-1G A695258-250MG
77293-234EA 19 USD
77293-234 77293-236
3-Amino-4-phenyl-1H-pyrazole 98%
3-Amino-4-phenyl-1H-pyrazole

Caution: for Research Use Only

Formula: C₉H₉N₃
MDL Number: MFCD01693715
CAS Number: 5591-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR