2-Amino-4-(4-fluorophenyl)thiazole 98%

Supplier: Ambeed
Warning

A686516-1G A686516-25G A686516-5G
77267-416EA 15 USD
77267-416 77267-418 77267-420
2-Amino-4-(4-fluorophenyl)thiazole 98%
2-Amino-4-(4-fluorophenyl)thiazole

Caution: for Research Use Only

Formula: C₉H₇FN₂S
MW: 194.2287 g/mol
MDL Number: MFCD01086598
CAS Number: 77815-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR