3,5-Bis(trifluoromethyl)phenyl isothiocyanate 95%

Supplier: Ambeed
Danger

Synonyms: 1,3-Bis(trifluoromethyl)-5-isothiocyanatobenzene, 1-Isothiocyanato-3,5-bis(trifluoromethyl)benzene

A661180-1G A661180-5G A661180-25G
77267-228EA 14 USD
77267-228 77267-230 77552-020
3,5-Bis(trifluoromethyl)phenyl isothiocyanate 95%
3,5-Bis(trifluoromethyl)phenyl isothiocyanate

Caution: for Research Use Only

Formula: C₉H₃F₆NS
MW: 271.19 g/mol
MDL Number: MFCD00040838
CAS Number: 23165-29-9
UN: 2206
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR