1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(+)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)benzene, (S,S)-Me-DUPHOS, 1,2-Bis((2S,5S)-2,5-dimethylphospholan-1-yl)benzene
A565684-100MG
A565684-1G
A565684-5G
A565684-250MG
77254-002EA
67
USD
77254-002
77254-004
77254-006
77572-798
1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene 98%
1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene
Caution: for Research Use Only
Formula:
C₁₈H₂₈P₂ |
MDL Number:
MFCD00142322 CAS Number: 136735-95-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...