2-Hydrazinoquinoline 98%

Supplier: Ambeed
Warning

Synonyms: 2-Quinolylhydrazine, 2-Hydrazinylquinoline

A562757-100MG A562757-1G A562757-250MG A562757-5G
77280-056EA 27 USD
77280-056 77280-058 77280-060 77280-062
2-Hydrazinoquinoline 98%
2-Hydrazinoquinoline

Caution: for Research Use Only

Formula: C₉H₉N₃
MW: 159.19 g/mol
MDL Number: MFCD00041860
CAS Number: 15793-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR