2-Aminoethyl hydrogen sulfate 95%

Supplier: Ambeed
Warning

Synonyms: 2-Aminoethyl Hydrogen sulfate, Sulfuric acid Mono(2-aminoethyl) Ester, 2-Aminoethyl Hydrogen sulphate

A536432-100G A536432-10G A536432-25G A536432-5G
77266-052EA 40 USD
77266-052 77266-054 77266-056 77266-058
2-Aminoethyl hydrogen sulfate 95%
2-Aminoethyl hydrogen sulphate

Caution: for Research Use Only

Formula: C₂H₇NO₄S
MDL Number: MFCD00008179
CAS Number: 926-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR