Adenine sulfate 97%

Supplier: Ambeed
Warning

Synonyms: 6-Aminopurine hemisulphate salt, 7H-Purin-6-amine hemisulphate, Adenine hemisulphate salt, 7H-Purin-6-amine hemisulfate, 6-Aminopurine hemisulfate salt, Adenine hemisulfate salt

A535802-100G A535802-25G A535802-500G
77279-912EA 52 USD
77279-912 77279-914 77749-118
Adenine sulfate 97%
Adenine sulphate

Caution: for Research Use Only

Formula: C₅H₅N₅·½H₂SO₄
MW: 184.17 g/mol
MDL Number: MFCD00213655
CAS Number: 321-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR