2-Ethylhexyl salicylate 95%
Supplier: Ambeed
|
Warning
|
A511686-1000G
A511686-100G
A511686-25G
A511686-500G
77265-808EA
112
USD
77265-808
77265-810
77265-812
77265-814
2-Ethylhexyl salicylate 95%
2-Ethylhexyl salicylate
Caution: for Research Use Only
Formula:
C₁₅H₂₂O₃ |
MDL Number:
MFCD00053300 CAS Number: 118-60-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...