4-Allylanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Allyl-4-methoxybenzene, Estragole

A509452-100G A509452-500G A509452-25G
77292-176EA 42 USD
77292-176 77292-178 77568-994
4-Allylanisole 98%
4-Allylanisole

Caution: For research use only.

Formula: C₁₀H₁₂O
MDL Number: MFCD00008653
CAS Number: 140-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR