α,2,6-Trichlorotoluene 98%

Supplier: Ambeed
Danger

Synonyms: 2,6-Dichlorobenzyl Chloride1,3-Dichloro-2-(chloromethyl)benzene2,6-Dichlorobenzyl chloride1-(Chloromethyl)-2,6-dichlorobenzene

A501530-5G A501530-25G
77253-304EA 15.45 USD
77253-304 77253-306
α,2,6-Trichlorotoluene 98%
α,2,6-Trichlorotoluene

Caution: for Research Use Only

Formula: C₇H₅Cl₃
MW: 195.47 g/mol
MDL Number: MFCD00000897
CAS Number: 2014-83-7
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR