2,4,6-Trifluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2,4,6-Trifluorophenyl)metaboric acid, 2,4,6-Trifluorobenzeneboronic acid, Dihydroxy(2,4,6-trifluorophenyl)borane

A500473-1G A500473-5G
77279-684EA 7 USD
77279-684 77279-686
2,4,6-Trifluorophenylboronic acid 98%
2,4,6-Trifluorophenylboronic acid

Caution: for Research Use Only

Formula: C₆H₄BF₃O₂
MDL Number: MFCD01863169
CAS Number: 182482-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR