α-Bromo-3-chlorotoulene 98%

Supplier: Ambeed
Danger

Synonyms: α-Bromo-m-chlorotoulene, 3-Chlorobenzyl bromide, 1-(Bromomethyl)-3-chlorobenzene, alpha-Bromo-3-chlorotoluene

A485240-10G A485240-25G A485240-5G
77279-540EA 18 USD
77279-540 77279-542 77279-544
α-Bromo-3-chlorotoulene 98%
α-Bromo-3-chlorotoulene

Caution: for Research Use Only

Formula: C₇H₆BrCl
MW: 205.48
Boiling Pt: 238
Melting Pt: 17.5
Density: 1.56
MDL Number: MFCD00000597
CAS Number: 766-80-3
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR