2,4,6-Tri(4-pyridyl)-1,3,5-triazine 95%

Supplier: Ambeed
Warning

A478339-1G A478339-250MG A478339-25G A478339-5G
77253-108EA 27 USD
77253-108 77253-110 77253-112 77253-114
2,4,6-Tri(4-pyridyl)-1,3,5-triazine 95%
2,4,6-Tri(4-pyridyl)-1,3,5-triazine

Caution: for Research Use Only

Formula: C₁₈H₁₂N₆
MDL Number: MFCD00129048
CAS Number: 42333-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR