Allyl phenyl sulfone 98%

Supplier: Ambeed
Warning

Synonyms: Phenyl 2-propenyl sulfone, Allyl Phenyl sulfone, Allylsulfonylbenzene, Allyl Phenyl sulphone, (Allylsulphonyl)benzene, (2-Propen-1-ylsulfonyl)benzene, (2-Propenylsulfonyl)benzene, (Allylsulfonyl)benzene

A446984-1G A446984-250MG
77252-706EA 26 USD
77252-706 77252-708
Allyl phenyl sulfone 98%
Allyl phenyl sulphone

Caution: for Research Use Only

Formula: C₉H₁₀O₂S
MDL Number: MFCD00014739
CAS Number: 16212-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR