1-Heptanol 95%

Supplier: Ambeed
Warning

A410251-100G A410251-500G A410251-1000G
77240-734EA 7 USD
77240-734 77240-736 77746-828
1-Heptanol 95%
1-Heptanol

Caution: for Research Use Only

Formula: H₃C(CH₂)₅CH₂OH
MDL Number: MFCD00002986
CAS Number: 111-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR