4-Dimethylaminoazobenzene-4'-carboxylic acid 97%

Supplier: Ambeed
Warning

A389198-100G A389198-1G A389198-25G A389198-5G
77291-046EA 544 USD
77291-046 77291-048 77291-050 77291-052
4-Dimethylaminoazobenzene-4'-carboxylic acid 97%
4-Dimethylaminoazobenzene-4'-carboxylic acid

Caution: for Research Use Only

Formula: C₁₅H₁₅N₃O₂
MDL Number: MFCD00058988
CAS Number: 6268-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR