6-Aminophthalide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Aminoisobenzofuran-1(3H)-one, 6-AMINOPHTHALIDE
A385893-100MG
A385893-250MG
A385893-1G
A385893-5G
77240-460EA
10
USD
77240-460
77240-462
77568-780
77572-148
6-Aminophthalide 98%
6-Aminophthalide
Caution: For research use only.
Formula:
C₈H₇NO₂ MW: 149.15 g/mol |
MDL Number:
MFCD00033530 CAS Number: 57319-65-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...