3,3',4,4'-Benzophenonetetracarboxylic dianhydride 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
Benzophenone-3,3',4,4'-tetracarboxylic dianhydride
A380623-100G
A380623-25G
A380623-500G
77278-526EA
36
USD
77278-526
77278-528
77278-530
3,3',4,4'-Benzophenonetetracarboxylic dianhydride 96%
3,3',4,4'-Benzophenonetetracarboxylic dianhydride
Caution: for Research Use Only
Formula:
C₁₇H₆O₇ MW: 322.23 g/mol |
MDL Number:
MFCD00005923 CAS Number: 2421-28-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...