(Benzoylmethylene)triphenylphosphorane 97%

Supplier: Ambeed
Warning

A376951-100G A376951-1G A376951-25G A376951-5G
77290-916EA 99 USD
77290-916 77290-918 77290-920 77290-922
(Benzoylmethylene)triphenylphosphorane 97%
(Benzoylmethylene)triphenylphosphorane

Caution: for Research Use Only

Formula: C₂₆H₂₁OP
MDL Number: MFCD00014088
CAS Number: 859-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR