4-Chlorobutyryl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 4-Chlorobutanoyl Chloride

A331098-25G A331098-100G
77290-448EA 24 USD
77290-448 77593-956
4-Chlorobutyryl chloride 98%
4-Chlorobutyryl chloride

Caution: for Research Use Only

Formula: C₄H₆Cl₂O
MDL Number: MFCD00000754
CAS Number: 4635-59-0
UN: 2922
ADR: 8,6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR