2-Bromo-4,6-diphenyl-1,3,5-triazine 97%

Supplier: Ambeed
Warning

A305884-1G A305884-5G
77263-108EA 21.99 USD
77263-108 77263-110
2-Bromo-4,6-diphenyl-1,3,5-triazine 97%
2-Bromo-4,6-diphenyl-1,3,5-triazine

Caution: for Research Use Only

Formula: C₁₅H₁₀BrN₃
MDL Number: MFCD10687147
CAS Number: 80984-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR