4-Chloro-2,6-diaminopyrimidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Chloropyrimidine-2,4-diamine
, 2,4-Diamino-6-chloropyrimidine
A302334-25G
A302334-5G
A302334-100G
A302334-1000G
A302334-500G
A302334-10G
77277-350EA
11
USD
77277-350
77277-354
77277-352
77277-346
77277-344
77277-348
4-Chloro-2,6-diaminopyrimidine 98%
4-Chloro-2,6-diaminopyrimidine
Caution: for Research Use Only
Formula:
C₄H₅ClN₄ MW: 144.56 g/mol |
MDL Number:
MFCD00006097 CAS Number: 156-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...