2-Amino-4,5-dimethylthiazole hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4,5-Dimethylthiazol-2-amine hydrochloride

A297065-100G A297065-25G A297065-5G
77250-758EA 56 USD
77250-758 77250-760 77250-762
2-Amino-4,5-dimethylthiazole hydrochloride 97%
2-Amino-4,5-dimethylthiazole hydrochloride

Caution: for Research Use Only

Formula: C₅H₉ClN₂S
MDL Number: MFCD00012711
CAS Number: 71574-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR