2-(Trifluoromethoxy)benzyl alcohol 97%

Supplier: Ambeed
Warning

A285083-1G A285083-25G
77238-962EA 12 USD
77238-962 77238-964
2-(Trifluoromethoxy)benzyl alcohol 97%
2-(Trifluoromethoxy)benzyl alcohol

Caution: for Research Use Only

Formula: C₈H₇F₃O₂
MDL Number: MFCD00153285
CAS Number: 175278-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR