2-Chlorobenzenesulfonamide 98%

Supplier: Ambeed
Warning

A284189-1000G A284189-100G A284189-25G A284189-500G
77289-690EA 135 USD
77289-690 77289-692 77289-694 77289-696
2-Chlorobenzenesulfonamide 98%
2-Chlorobenzenesulphonamide

Caution: for Research Use Only

Formula: C₆H₆ClNO₂S
MDL Number: MFCD00051974
CAS Number: 6961-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR