2-Acetyl-4-methylthiophene 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Methyl-2-thienyl)-1-ethanone, 1-(4-Methylthiophen-2-yl)ethanone

A277870-100MG A277870-250MG
77250-466EA 8 USD
77250-466 77250-468
2-Acetyl-4-methylthiophene 98%
2-Acetyl-4-methylthiophene

Caution: for Research Use Only

Formula: C₇H₈OS
MW: 140.21 g/mol
MDL Number: MFCD00027395
CAS Number: 13679-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR