NSC697923 97%

Supplier: Ambeed
Danger

A274860-1MG A274860-25MG A274860-5MG
77238-800EA 30 USD
77238-800 77238-802 77238-804
NSC697923 97%
NSC697923

Caution: for Research Use Only

Formula: C₁₁H₉NO₅S
MDL Number: MFCD01925132
CAS Number: 343351-67-7
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR