4-Fluoro-DL-phenylglycine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-DL-α-phenylglycine, 4-Fluorophenylglycine, H-DL-Phg(4-F)-OH

A268700-10G A268700-1G A268700-5G
77250-296EA 25 USD
77250-296 77250-298 77250-300
4-Fluoro-DL-phenylglycine 97%
4-Fluoro-DL-phenylglycine

Caution: for Research Use Only

Formula: C₈H₈FNO₂
MW: 169.155 g/mol
MDL Number: MFCD00066444
CAS Number: 7292-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR