2'-Deoxy-5-fluorocytidine 95%

Supplier: Ambeed
Warning

A266441-100MG A266441-1MG
77262-354EA 15 USD
77262-354 77262-356
2'-Deoxy-5-fluorocytidine 95%
2'-Deoxy-5-fluorocytidine

Caution: for Research Use Only

Formula: C₉H₁₂FN₃O₄
MDL Number: MFCD00077348
CAS Number: 10356-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR