2-Amino-4-chloro-6-isopropylpyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-6-isopropylpyrimidin-2-amine

A257710-100MG A257710-1G A257710-250MG A257710-5G
77250-090EA 5 USD
77250-090 77250-092 77250-094 77250-096
2-Amino-4-chloro-6-isopropylpyrimidine 98%
2-Amino-4-chloro-6-isopropylpyrimidine

Caution: for Research Use Only

Formula: C₇H₁₀ClN₃
MDL Number: MFCD03840198
CAS Number: 73576-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR