8-Azaadenine 98%

Supplier: Ambeed
Warning

A254981-100MG A254981-1G A254981-250MG
77289-126EA 41.2 USD
77289-126 77289-128 77289-130
8-Azaadenine 98%
8-Azaadenine

Caution: for Research Use Only

Formula: C₄H₄N₆
MDL Number: MFCD00005697
CAS Number: 1123-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR