SMER28 98%

Supplier: Ambeed
Warning

Synonyms: N-Allyl-6-bromoquinazolin-4-amine

A249882-1MG A249882-25MG A249882-5MG
77238-260EA 60.77 USD
77238-260 77238-262 77238-264 77238-266
SMER28 98%
SMER28

Caution: for Research Use Only

Formula: C₁₁H₁₀BrN₃
MDL Number: MFCD02166825
CAS Number: 307538-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR