2-Iodophenylboronic acid 98%

Supplier: Ambeed
Warning

A245675-100MG A245675-1G
77276-166EA 15 USD
77276-166 77276-168
2-Iodophenylboronic acid 98%
2-Iodophenylboronic acid

Caution: for Research Use Only

Formula: C₆H₆BIO₂
MDL Number: MFCD09743934
CAS Number: 1008106-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR