2-Amino-1,3,4-thiadiazole 97%

Supplier: Ambeed
Warning

A232255-1000G A232255-100G A232255-1G A232255-25G A232255-500G
77288-438EA 139 USD
77288-438 77288-440 77288-442 77288-444 77288-446 77288-448
2-Amino-1,3,4-thiadiazole 97%
2-Amino-1,3,4-thiadiazole

Caution: for Research Use Only

Formula: C₂H₃N₃S
MDL Number: MFCD00003107
CAS Number: 4005-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR